Order Entry
VWR®, LTS® Compatible Filter Tips
undefined
VWR®, LTS® Compatible Filter Tips
  • Catalog No:
  • 732-3641
  • 732-3642
  • 732-3643
  • 732-3639
  • 732-3640
  • Environmentally Preferable:
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Waste Reducing, Sustainable Packaging
  • Environmental certification:
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  • FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
  •  
    Volume
    Sterile
    Filter
    Length
    Colour code
    For use with
    Packed
  • 732-3639
    20 µl
    +
    +
    49,63 mm
    Red
    Rainin LTS style pipettes
    10 racks, 96 each
    1 * 960 items
    1 * 960 items
      Waste Reducing
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Waste Reducing, Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing
      Sustainable Packaging
  • 732-3640
    200 µl
    +
    +
    50,75 mm
    Green
    Rainin LTS style pipettes
    10 racks, 96 each
    1 * 960 items
    1 * 960 items
      Waste Reducing
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Waste Reducing, Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing
      Sustainable Packaging
  • 732-3641
    300 µl
    +
    +
    58,93 mm
    Yellow
    Rainin LTS style pipettes
    10 racks, 96 each
    1 * 960 items
    1 * 960 items
      Waste Reducing
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Waste Reducing, Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing
      Sustainable Packaging
  • 732-3642
    1000 µl
    +
    +
    88,90 mm
    Blue
    Rainin LTS style pipettes
    8 racks, 96 each
    1 * 768 items
    1 * 768 items
      Waste Reducing
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Waste Reducing, Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing
      Sustainable Packaging
  • 732-3643
    1200 µl
    +
    +
    97,66 mm
    Grey
    Rainin LTS style pipettes
    8 racks, 96 each
    1 * 768 items
    1 * 768 items
      Waste Reducing
      Sustainable Packaging
    Specifications
    Environmentally Preferable
    Waste Reducing, Sustainable Packaging
    Environmental certification
    FSC-STD-40-004 V3-1 and FSC-C154485; Compared to competitor racks, these racks use 1/3 less plastic
      Waste Reducing
      Sustainable Packaging