Digital Solutions
Applications
Support
About
Order Entry
VWR®, Viscosity check oils
:
VWR Chemicals
VWR®, Viscosity check oils
:
VWR Chemicals
SDS_EN_UK_VWRC99171.pdf
SDS_EN_UK_VWRC99172.pdf
VWR® for Calibrations
Viscosity at 40 °C
Pk
Check Oil 10W30
Check Oil 10W30
99171.260
67,27 mm²/s
500 mL
1 * 500 mL
-
+
1 * 500 mL
-
+
Check Oil 10W30
Check Oil 10W30
99171.360
67,27 mm²/s
5 L
1 * 5 L
-
+
1 * 5 L
-
+
Check Oil 15W40
Check Oil 15W40
99172.260
103,1 mm²/s
500 mL
1 * 500 mL
-
+
1 * 500 mL
-
+
Check Oil 15W40
Check Oil 15W40
99172.360
103,1 mm²/s
5 L
1 * 5 L
-
+
1 * 5 L
-
+
Check Oil 5W30
Check Oil 5W30
99170.260
65,90 mm²/s
500 mL
1 * 500 mL
-
+
1 * 500 mL