Order Entry
VWR® ARISTAR®, Molybdenum Standard Solution, 1000 mg/L Mo in Ammonium Hydroxide Solution (max. 1%) (from (NH₄)₂MoO₄), Standard for ICP
undefined
VWR® ARISTAR®, Molybdenum Standard Solution, 1000 mg/L Mo in Ammonium Hydroxide Solution (max. 1%) (from (NH₄)₂MoO₄), Standard for ICP
  •  
    Element
    Symbol
    Concentration
    Matrix
    Pack type
    Pk
  • 455684P
    Molybdenum
    Mo
    1000 mg/L
    <1% NH₄OH
    Plastic bottle
    500 mL
    1 * 500 mL
    1 * 500 mL
  • 455682N
    Molybdenum
    Mo
    1000 mg/L
    <1% NH₄OH
    Plastic bottle
    100 mL
    1 * 100 mL
    1 * 100 mL