Digital Solutions
Applications
Support
About
$0.00
Order Entry
VWR®, Residual solvent standards, Class 3
:
VWR Chemicals
VWR®, Residual solvent standards, Class 3
:
VWR Chemicals
SDS_EN_UK_VWRC88210.pdf
sds_VWRC88210
sds_VWRC88213
VWR® for Calibrations
Concentration
Standards
Pk
Class III Mix A (23 components)
Class III Mix A (23 components)
88210.001
5000 μg/ml
Residual Solvent
1 mL
1 * 1 mL
-
+
1 * 1 mL
-
+
Class III Mix B (2 components)
Class III Mix B (2 components)
88211.001
5000 μg/ml
Residual Solvent
1 mL
1 * 1 mL
-
+
1 * 1 mL
-
+
Class III Mix C (1 component)
Class III Mix C (1 component)
88213.001
5000 μg/ml
Residual Solvent
1 mL
1 * 1 mL
-
+
1 * 1 mL