About this item
High-performance pump tubing for challenging applications.
- Genuine Masterflex® tubing undergoes robust testing to provide optimal performance / pump life hours with peristaltic pump equipment
- Precision extruded to tight tolerances for high repeatability, accuracy and reproducibility
- Global market leader in range of formulations for most extensive chemical compatibility
- Single-use custom lengths and configurations available to minimize downtime
- Regulatory approvals to match your unique application
- Custom extruded to fit Masterflex® pumps
- Lot-to-lot consistency provides superior accuracy and repeatability
Applications: Chemical and pharmaceutical applications
Characteristics: Enhanced chemical resistance, including aggressive organic solvents, spallation-free, low gas permeability, opaque white
Gore® STA-PURE® PFL-Series Pump Tubing is your ideal choice for high purity or long-running, critical pharmaceutical and biopharmaceutical processing applications. The patented composite (perfluoroelastomer) and ePTFE (polytetrafluoroethylene), results in a tube that offers high flex durability for greater resistance to rupture—even at applications up to 60 psig. Long-term pump testing proves that Gore® STA-PURE® PFL-Series Pump Tubing lasts hundreds of hours with consistent and repeatable flow rates resulting in one saving time and reducing maintenance by having fewer tubing changes.
If one is looking for a flow stable / process stable tubing that withstands CIP (clean-in-place) or SIP (steam-in-place) cleaning—look no further than this tubing!
Gore® STA-PURE® Pump tubing - Series PFL is available as single tube elements. Recommended for use in L/S® and I/P® standard and Easy-Load® pump heads, and L/S® Easy-Load® II pump heads.
NOTE: There is an engineering questionnaire vetting application specific details before the order can be processed.
For custom tubing lengths and configurations, please contact us. We can provide the tubing type, length, and quantity you need.
- Environmentally Preferable:
Specifications
- Catalog No:
- MFLX96242-24
- MFLX96242-25
- MFLX96242-17
- MFLX96242-18
- MFLX96242-35
- MFLX96242-36
- MFLX96242-15
- MFLX96242-16
- MFLX96242-14
- Model:
- 96242-24
- 96242-25
- 96242-17
- 96242-18
- 96242-35
- 96242-36
- 96242-15
- 96242-16
- 96242-14
- Length per pack:
- 0,33 m (1,1 ft.)
- 0,35 m (1 ft.)
- 0,35 m (1 ft.)
- 0,35 m (1 ft.)
- 0,33 m (1,1 ft.)
- 0,33 m (1,1 ft.)
- 0,33 m (1,1 ft.)
- 0,35 m (1 ft.)
- 0,35 m (1 ft.)
- Max. pressure:
- 4,1 bar (60 psi)
- 4,1 bar (60 psi)
- 4,1 bar (60 psi)
- 4,1 bar (60 psi)
- 4,1 bar (60 psi)
- 6,8 bar (100 psi)
- 4,1 bar (60 psi)
- 4,1 bar (60 psi)
- 4,1 bar (60 psi)
- Min. temperature:
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- –80 °C (–112 °F)
- Max. temperature:
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- 200 °C (392 °F)
- Min. temperature (dynamic):
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- –40 °C (–40 °F)
- Max. temperature (dynamic):
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- 150 °C (302 °F)
- Max. vacuum:
- 660 mm Hg (26" Hg)
- 660 mm Hg (26" Hg)
- 510 mm Hg (20" Hg)
- 510 mm Hg (20" Hg)
- 660 mm Hg (26" Hg)
- 610 mm Hg (24" Hg)
- 660 mm Hg (26" Hg)
- 660 mm Hg (26" Hg)
- 660 mm Hg (26" Hg)
- Pump series:
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- L/S
- Regulatory approvals:
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- ADCF;Non-DEHP;RoHS;USP
- Sterilisation:
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Autoclave, SIP
- Sterilisation method autoclave:
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Yes
- Sterilisation method EtO:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Sterilisation method gamma:
- No
- No
- No
- No
- No
- No
- No
- No
- No
- Suction lift H₂O:
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- 6,7 m (22 ft.)
- 6,7 m (22 ft.)
- 8,8 m (29 ft.)
- 8,3 m (27 ft.)
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- 8,8 m (29 ft.)
- Warranty:
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
- Out of box
Specifications
Related Information
Product Information
- Catalog #Tubing int.ØTubing materialL/S Masterflex tubing sizeHose barb sizeMaximum Flow RatePkAvailabilityPrice
- L/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 14; 12"/PkL/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 14; 12"/Pk1,60 mm (0,063")GORE® STA-PURE®140,0625"130 ml/min12 "Log in for availability1 * 12 "Specifications:CE certificate
More
Specifications
Model96242-14Environmentally PreferableLength per pack0,35 m (1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,8 m (29 ft.)WarrantyOut of boxCE certificate - L/S® High-Performance Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 15; 12"/PkL/S® High-Performance Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 15; 12"/Pk4,80 mm (0,189")GORE® STA-PURE®150,1875"1100 ml/min12 "Log in for availability1 * 12 "Specifications:CE certificate
More
Specifications
Model96242-15Environmentally PreferableLength per pack0,33 m (1,1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,8 m (29 ft.)WarrantyOut of boxCE certificate - L/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 16; 12"/PkL/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 16; 12"/Pk3,10 mm (0,122")GORE® STA-PURE®160,125"480 ml/min12 "Log in for availability1 * 12 "Specifications:CE certificate
More
Specifications
Model96242-16Environmentally PreferableLength per pack0,35 m (1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,8 m (29 ft.)WarrantyOut of boxCE certificate - L/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 17; 12"/PkL/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 17; 12"/Pk6,40 mm (0,252")GORE® STA-PURE®170,25"1700 ml/min12 "Log in for availability1 * 12 "Specifications:CE certificate
More
Specifications
Model96242-17Environmentally PreferableLength per pack0,35 m (1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum510 mm Hg (20" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O6,7 m (22 ft.)WarrantyOut of boxCE certificate - L/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 18; 12"/PkL/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 18; 12"/Pk7,90 mm (0,311")GORE® STA-PURE®180,375"2300 ml/min12 "Log in for availability1 * 12 "Specifications:CE certificate
More
Specifications
Model96242-18Environmentally PreferableLength per pack0,35 m (1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum510 mm Hg (20" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O6,7 m (22 ft.)WarrantyOut of boxCE certificate - L/S® High-Performance Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 24; 14"/PkL/S® High-Performance Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 24; 14"/Pk6,40 mm (0,252")GORE® STA-PURE®240,25"1800 ml/min14 "Log in for availability1 * 14 "Specifications:CE certificate
More
Specifications
Model96242-24Environmentally PreferableLength per pack0,33 m (1,1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,8 m (29 ft.)WarrantyOut of boxCE certificate - L/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 25; 12"/PkL/S® Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 25; 12"/Pk4,80 mm (0,189")GORE® STA-PURE®250,1875"1000 ml/min12 "Log in for availability1 * 12 "Specifications:CE certificate
More
Specifications
Model96242-25Environmentally PreferableLength per pack0,35 m (1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,8 m (29 ft.)WarrantyOut of boxCE certificate - L/S® High-Performance Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 35; 14"/PkL/S® High-Performance Precision Pump Tubing, Gore® STA-PURE® PFL-Series, L/S 35; 14"/Pk7,90 mm (0,311")GORE® STA-PURE®350,375"2600 ml/min14 "Log in for availability1 * 14 "Specifications:CE certificate
More
Specifications
Model96242-35Environmentally PreferableLength per pack0,33 m (1,1 ft.)Max. pressure4,1 bar (60 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum660 mm Hg (26" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,8 m (29 ft.)WarrantyOut of boxCE certificate - L/S 36 Gore Sta-Pure Pump Tubing, PFL-Series; 14"/PkL/S 36 Gore Sta-Pure Pump Tubing, PFL-Series; 14"/Pk9,70 mm (0,382")GORE® STA-PURE®360,375"3400 ml/min14 "Log in for availability1 * 14 "Specifications:CE certificate
More
Specifications
Model96242-36Environmentally PreferableLength per pack0,33 m (1,1 ft.)Max. pressure6,8 bar (100 psi)Min. temperature–80 °C (–112 °F)Max. temperature200 °C (392 °F)Min. temperature (dynamic)–40 °C (–40 °F)Max. temperature (dynamic)150 °C (302 °F)Max. vacuum610 mm Hg (24" Hg)Pump seriesL/SRegulatory approvalsADCF;Non-DEHP;RoHS;USPSterilisationAutoclave, SIPSterilisation method autoclaveYesSterilisation method EtONoSterilisation method gammaNoSuction lift H₂O8,3 m (27 ft.)WarrantyOut of boxCE certificate