Order Entry
VWR®, Dispensers for Personal Protective Equipment
VWR®, Dispensers for Personal Protective Equipment
Supplier:  VWR Collection
Certificates
undefined
VWR®, Dispensers for Personal Protective Equipment
Specifications
  • Catalog No:
  • 111-0000
  • 111-0003
  • 111-0019
  • 111-0002
  • 111-0022
  • 111-0010
  • 111-0021
  • 111-0015
  • 111-0163
  • 111-0013
  • 111-0161
  • 111-0025
  • 111-0014
  • 111-0162
  • PPE type:
  • Safety glasses
  • Helmets
  • Hairnets
  • Safety glasses
  • Respirators
  • Hairnets
  • Safety glasses
  • Respirators
  • Helmets
  • Gloves
  • Earplugs
  • Gloves
  • Hairnets
  • Hairnets
  • Material:
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • ABS
  • Dispenser type:
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • Wall-mountable
  • W×D×H:
  • 236×120×120 mm
  • 236×200×315 mm
  • 236×125×225 mm
  • 236×125×225 mm
  • 236×125×225 mm
  • 236×120×120 mm
  • 236×120×120 mm
  • 236×125×225 mm
  • 236×200×315 mm
  • 236×120×120 mm
  • 236×125×225 mm
  • 236×120×120 mm
  • 236×120×120 mm
  • 236×125×225 mm
Specifications
Mini
Product Information
  •  
    Colour
    W×D×H
    Pk
  • Safety glasses dispenser, ABS
    Safety glasses dispenser, ABS
    111-0000
    Blue
    236×120×120 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Safety glasses
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×120×120 mm
  • Safety glasses dispenser, ABS
    Safety glasses dispenser, ABS
    111-0021
    Transparent
    236×120×120 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Safety glasses
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×120×120 mm
  • Hairnet dispenser, ABS
    Hairnet dispenser, ABS
    111-0014
    Blue
    236×120×120 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Hairnets
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×120×120 mm
  • Hairnet dispenser, ABS
    Hairnet dispenser, ABS
    111-0010
    Transparent
    236×120×120 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Hairnets
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×120×120 mm
  • Hand protection, ABS
    Hand protection, ABS
    111-0013
    Transparent
    236×120×120 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Gloves
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×120×120 mm
  • Hand protection dispenser, ABS
    Hand protection dispenser, ABS
    111-0025
    Blue
    236×120×120 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Gloves
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×120×120 mm
Midi
  •  
    Colour
    W×D×H
    Pk
  • Ear protection dispenser, ABS
    Ear protection dispenser, ABS
    111-0002
    Blue
    236×125×225 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Safety glasses
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×125×225 mm
  • Respirator dispenser, ABS
    Respirator dispenser, ABS
    111-0022
    Blue
    236×125×225 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Respirators
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×125×225 mm
  • Hairnet dispenser, ABS
    Hairnet dispenser, ABS
    111-0019
    Blue
    236×125×225 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Hairnets
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×125×225 mm
  • Respirator dispenser, ABS
    Respirator dispenser, ABS
    111-0015
    Transparent
    236×125×225 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Respirators
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×125×225 mm
  • Ear protection dispenser, ABS
    Ear protection dispenser, ABS
    111-0161
    Transparent
    236×125×225 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Earplugs
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×125×225 mm
  • Hairnet dispenser, ABS
    Hairnet dispenser, ABS
    111-0162
    Transparent
    236×125×225 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Hairnets
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×125×225 mm
Maxi
  •  
    Colour
    W×D×H
    Pk
  • Helmet dispenser, ABS
    Helmet dispenser, ABS
    111-0003
    Blue
    236×200×315 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Helmets
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×200×315 mm
  • Helmet and safety glasses dispenser, ABS
    Helmet and safety glasses dispenser, ABS
    111-0163
    Blue
    236×200×315 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    PPE type
    Helmets
    Material
    ABS
    Environmentally Preferable
    Dispenser type
    Wall-mountable
    W×D×H
    236×200×315 mm