Order Entry
Accessories for VWR® Mini Blot Shaker
undefined
Accessories for VWR® Mini Blot Shaker
  •  
    Pk
  • Large blot box, 117×89 mm
    Large blot box, 117×89 mm
    700-0248
    10
    1 * 10 items
    1 * 10 items
    Specifications
    Environmentally Preferable
  • Small blot box, 91×66 mm
    Small blot box, 91×66 mm
    700-0249
    10
    1 * 10 items
    1 * 10 items
    Specifications
    Environmentally Preferable