Order Entry
VWR® Cryoboxes and Dividers
undefined
VWR® Cryoboxes and Dividers
Mechanical Cryogenic Freezer Boxes
  •  
    Compartments
    Colour
    W×D×H ext.
    Pk
  • Cryobox without divider, waterproof
    Cryobox without divider, waterproof
    479-2220
    White
    127×127×73,0 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Cryobox without divider, waterproof
    Cryobox without divider, waterproof
    479-2215
    White
    133×133×50 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Cryobox with divider (height 30 mm), waterproof, with holes in the bottom
    Cryobox with divider (height 30 mm), waterproof, with holes in the bottom
    479-2217
    25 (5×5)
    White
    75×75×50 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Cryobox with divider (height 30 mm), waterproof, with holes in the bottom
    Cryobox with divider (height 30 mm), waterproof, with holes in the bottom
    479-2213
    81 (9×9)
    White
    133×133×50 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Cryobox with divider (height 20 mm), water repellent, for 0,2 ml PCR tubes
    Cryobox with divider (height 20 mm), water repellent, for 0,2 ml PCR tubes
    479-2227
    81 (9×9)
    White
    76×76×24,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • Cryobox with divider (height 20 mm), water repellent, for 0,2 ml PCR tubes
    Cryobox with divider (height 20 mm), water repellent, for 0,2 ml PCR tubes
    479-2228
    196 (14×14)
    White
    133×133×24,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
Fiberboard Dividers
  •  
    Compartments
    Colour
    W×D×H ext.
    Pk
  • VWRU89214-740
    89214-740
    25 (for mini bo×)
    67×67×32 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • 479-2224
    -
    25
    133x133x 25,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • 479-2223
    -
    49
    125×125×25,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • VWRU82021-122
    82021-122
    49
    125×125×25,4 mm
    12
    1 * 12 items
    1 * 12 items
    Specifications
    Environmentally Preferable
  • 479-2219
    -
    64
    133x133x 25,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • VWRU82021-124
    82021-124
    64
    125×125×25,4 mm
    12
    1 * 12 items
    1 * 12 items
    Specifications
    Environmentally Preferable
  • 479-2221
    -
    81
    125×125×25,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • 479-2216
    -
    100
    133x133x25,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable
  • 479-2225
    -
    16
    125×125×25,4 mm
    1
    1 * 1 items
    1 * 1 items
    Specifications
    Environmentally Preferable