Order Entry
VWR® Ultra-High Performance, Multi-channel pipettes, mechanical, variable volume
VWR® Ultra-High Performance, Multi-channel pipettes, mechanical, variable volume
Supplier:  VWR Collection
Certificates
undefined
VWR® Ultra-High Performance, Multi-channel pipettes, mechanical, variable volume
Supplier:  VWR Collection
8-Channel
Product Information
  • Catalog #
    Volume
    Accuracy
    Imprecision
    Colour code
    Pk
    Availability
    Price
  • 613-7070
    0,5 - 10 µl
    ±10,0 - ±2,0 %
    ≤8,0 - ≤1,2 %
    Red
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7071
    5 - 50 µl
    ±4,0 - ±1,6 %
    ≤2,5 - ≤0,6 %
    Yellow
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7072
    10 - 200 µl
    ±3,0 - ±1,0 %
    ≤1,5 - ≤0,6 %
    Yellow
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7073
    30 - 300 µl
    ±3,0 - ±1,0 %
    ≤2,0 - ≤0,6 %
    Yellow
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
12-Channel
Product Information
  • Catalog #
    Volume
    Accuracy
    Imprecision
    Colour code
    Pk
    Availability
    Price
  • 613-7074
    0,5 - 10 µl
    ±10,0 - ±2,0 %
    ≤8,0 - ≤1,2 %
    Red
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7075
    5 - 50 µl
    ±4,0 - ±1,6 %
    ≤2,5 - ≤0,6 %
    Yellow
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7076
    10 - 200 µl
    ±3,0 - ±1,0 %
    ≤1,5 - ≤0,6 %
    Yellow
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7077
    30 - 300 µl
    ±1,6 - ±1,0 %
    ≤1,5 - ≤0,6 %
    Yellow
    1
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty