Order Entry
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Supplier:  VWR Collection
Certificates
undefined
VWR® Ultra-High Performance, Single Channel Pipettes, Mechanical, Variable Volume
Product Information
  • Catalog #
    Volume
    Accuracy
    Imprecision
    Colour code
    Pk
    Availability
    Price
  • 613-7065
    2 - 20 µl
    ±3,5 - ±0,8 %
    ≤1,5 - ≤0,3 %
    Yellow
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7063
    0,1 - 2 µl
    ±40,0 - ±1,5 %
    ≤12,0 - ≤0,7 %
    Red
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7064
    0,5 - 10 µl
    ±4,0 - ±0,5 %
    ≤2,8 - ≤0,4 %
    Red
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7066
    5 - 50 µl
    ±3,5 - ±0,8 %
    ≤2,0 - ≤0,4 %
    Yellow
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7067
    10 - 100 µl
    ±3,0 - ±0,8 %
    ≤1,0 - ≤0,2 %
    Yellow
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7068
    20 - 200 µl
    ±2,0 - ±0,6 %
    ≤0,7 - ≤0,2 %
    Yellow
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 613-7069
    100 - 1000 µl
    ±2,5 - ±0,6 %
    ≤0,6 - ≤0,2 %
    Blue
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
Starter Kit
Product Information
  • Catalog #
    Volume
    Accuracy
    Imprecision
    Colour code
    Pk
    Availability
    Price
  • 3 Pack incl. 3 pipettes (0,5 - 10 / 10 - 100 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and 8 pipette multiple stand
    3 Pack incl. 3 pipettes (0,5 - 10 / 10 - 100 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and 8 pipette multiple stand
    613-7078
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 3 Pack incl. 3 pipettes (2 - 20 / 20 - 200 / 100 - 1000 µl), 2 racks of tips (200 and 1000 µl) and 8 pipette multiple stand
    3 Pack incl. 3 pipettes (2 - 20 / 20 - 200 / 100 - 1000 µl), 2 racks of tips (200 and 1000 µl) and 8 pipette multiple stand
    613-7079
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 4 Pack incl. 4 pipettes (0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and 8 pipette multiple stand
    4 Pack incl. 4 pipettes (0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and 8 pipette multiple stand
    613-7080
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty
  • 5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and 8 pipette multiple stand
    5 Pack incl. 5 pipettes (0,1 - 2 / 0,5 - 10 / 2 - 20 / 20 - 200 / 100 - 1000 µl), 3 racks of tips (10, 200 and 1000 µl) and 8 pipette multiple stand
    613-7081
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    5 year warranty
    Accessories
    Specifications
    Environmentally Preferable
    5 year warranty