VWR® SuperFrost® BS 7011, Microscope Slides
Supplier: VWR Collection
Certificates
About this item
These microscope slides come with a light opaque coating in different colours.
- Resistant to commonly used solvents
- No scratching or sticking when stacked together
- Label areas on the top of the slide, for quick identification
Each colour codes different specimens at a glance according to urgency, processing or staining technique.
Certifications: BS 7011
Related Information
Certificates
Search by Lot NumberCoC Microscope Sildes, Superfrost® BS 7011, VWR® COC_VWRI631-0909_01322 COC_VWRI631-0909_02322 COC_VWRI631-0909_03323 COC_VWRI631-0909_04323
More
VWR® for Microscopy
Product Literature
VWR® Microscope Slides
Product specification
Product Information
- Catalog #ColourThicknessPkAvailabilityPrice
- Microscope slides (ground edges 90°)Microscope slides (ground edges 90°)631-0909White0,95 - 1,05 mm50Log in for availability1 * 50 itemsSpecifications:
Specifications
Environmentally Preferable - Microscope slides (ground edges 90°)Microscope slides (ground edges 90°)631-0910Blue0,95 - 1,05 mm50Log in for availability1 * 50 itemsSpecifications:
Specifications
Environmentally Preferable - Microscope slides (ground edges 90°)Microscope slides (ground edges 90°)631-0911Pink0,95 - 1,05 mm50Log in for availability1 * 50 itemsSpecifications:
Specifications
Environmentally Preferable - Microscope slides (ground edges 90°)Microscope slides (ground edges 90°)631-0912Yellow0,95 - 1,05 mm50Log in for availability1 * 50 itemsSpecifications:
Specifications
Environmentally Preferable - Microscope slides (ground edges 90°)Microscope slides (ground edges 90°)631-0913Green0,95 - 1,05 mm50Log in for availability1 * 50 itemsSpecifications:
Specifications
Environmentally Preferable