Order Entry
VWR®, Tris-borate-EDTA (TBE) buffer
undefined
VWR®, Tris-borate-EDTA (TBE) buffer
  •  
    Pack type
    Pk
  • TBE buffer, 10X liquid concentrate
    TBE buffer, 10X liquid concentrate
    0658-1L
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L
     : VWRC0658-1L
  • TBE buffer, 10X liquid concentrate
    TBE buffer, 10X liquid concentrate
    0658-4L
    Bag-in-box
    4 L
    1 * 4 L
    1 * 4 L
     : VWRC0658-4L
  • TBE buffer, 10X Ready-Pack™, each pack makes 1 L of 10X TBE buffer
    TBE buffer, 10X Ready-Pack™, each pack makes 1 L of 10X TBE buffer
    0478-2PK
    Pouch
    2
    1 * 2 items
    1 * 2 items
     : VWRC0478-2PK
  • TBE buffer, 10X solution
    TBE buffer, 10X solution
    0478-40L
    Plastic bottle for solids
    1 kg
    1 * 1 kg
    1 * 1 kg
     : VWRC0478-40L
  • TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure
    J885-4L
    Bag-in-box
    4 L
    1 * 4 L
    1 * 4 L
     : VWRCJ885-4L
  • TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure
    TBE buffer solution 5X concentrate (TRIS-borate-EDTA buffer), ultrapure
    J885-1L
    Plastic bottle
    1 L
    1 * 1 L
    1 * 1 L
     : VWRCJ885-1L