Order Entry
VWR®, Test Tubes Racks
VWR®, Test Tubes Racks
Supplier:  VWR Collection
Certificates
undefined
VWR®, Test Tubes Racks
Supplier:  VWR Collection
Certificates
Product Information
  • Catalog #
    Colour
    No. of Holes
    For tube Ø
    W×D×H
    Pk
    Availability
    Price
  • 212-0396
    Blue
    9 (3×3)
    30 mm
    110,6×110,6×84,2 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0397
    Green
    9 (3×3)
    30 mm
    110,6×110,6×84,2 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0398
    Red
    9 (3×3)
    30 mm
    110,6×110,6×84,2 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0399
    White
    9 (3×3)
    30 mm
    110,6×110,6×84,2 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0394
    Blue
    16 (4×4)
    25 mm
    123,9×123,9×74 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0395
    White
    16 (4×4)
    25 mm
    123,9×123,9×74 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0390
    Blue
    20 (4×5)
    20 mm
    130×105×84 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0391
    Green
    20 (4×5)
    20 mm
    130×105×84 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0392
    Red
    20 (4×5)
    20 mm
    130×105×84 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0393
    White
    20 (4×5)
    20 mm
    130×105×84 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0425
    Blue
    24 (3×8)
    30 mm
    110×285,5×86,7 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0426
    Green
    24 (3×8)
    30 mm
    110×285,5×86,7 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0427
    Orange
    24 (3×8)
    30 mm
    110×285,5×86,7 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0428
    Red
    24 (3×8)
    30 mm
    110×285,5×86,7 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0429
    White
    24 (3×8)
    30 mm
    110×285,5×86,7 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0430
    Yellow
    24 (3×8)
    30 mm
    110×285,5×86,7 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0382
    Blue
    36 (6×6)
    13 mm
    106×106×57 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0383
    Green
    36 (6×6)
    13 mm
    106×106×57 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0384
    Red
    36 (6×6)
    13 mm
    106×106×57 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0385
    White
    36 (6×6)
    13 mm
    106×106×57 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0386
    Blue
    36 (6×6)
    16 mm
    131×131×70 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0387
    Green
    36 (6×6)
    16 mm
    131×131×70 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0388
    Red
    36 (6×6)
    16 mm
    131×131×70 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0389
    White
    36 (6×6)
    16 mm
    131×131×70 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0413
    Blue
    40 (4×10)
    20 mm
    104×253,6×86,6 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0414
    Green
    40 (4×10)
    20 mm
    104×253,6×86,6 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0415
    Orange
    40 (4×10)
    20 mm
    104×253,6×86,6 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0416
    Red
    40 (4×10)
    20 mm
    104×253,6×86,6 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0417
    White
    40 (4×10)
    20 mm
    104×253,6×86,6 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0418
    Yellow
    40 (4×10)
    20 mm
    104×253,6×86,6 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0419
    Blue
    40 (4×10)
    25 mm
    123×301×94,5 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0420
    Green
    40 (4×10)
    25 mm
    123×301×94,5 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0421
    Orange
    40 (4×10)
    25 mm
    123×301×94,5 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0422
    Red
    40 (4×10)
    25 mm
    123×301×94,5 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0423
    White
    40 (4×10)
    25 mm
    123×301×94,5 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0424
    Yellow
    40 (4×10)
    25 mm
    123×301×94,5 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0400
    Blue
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0401
    Green
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0402
    Magenta
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0403
    Orange
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0404
    Red
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0405
    White
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0406
    Yellow
    72 (6×12)
    13 mm
    103,5×202,5×60 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0407
    Blue
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0408
    Green
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0409
    Orange
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0410
    Red
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0411
    White
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable
  • 212-0412
    Yellow
    72 (6×12)
    16 mm
    127,3×250,6×73 mm
    1
    Log in for availability
    1 * 1 items
    1 * 1 items
    Specifications:
    Specifications
    Environmentally Preferable