Order Entry
VWR®, Biohazard bags with or without sterilisation indicator strip
undefined
VWR®, Biohazard bags with or without sterilisation indicator strip
'Biobag' autoclavable bags
  •  
    Material
    L×W
    Pk
  • Autoclavable bags, resistant to 121 °C , 'Biobag' type 30
    Autoclavable bags, resistant to 121 °C , 'Biobag' type 30
    129-0714
    Plastic
    300×500 mm
    500
    1 * 500 items
    1 * 500 items
    Specifications
    Environmentally Preferable
  • Autoclavable bags, resistant to 121 °C , 'Biobag' type 60
    Autoclavable bags, resistant to 121 °C , 'Biobag' type 60
    129-0715
    Plastic
    600×780 mm
    500
    1 * 500 items
    1 * 500 items
    Specifications
    Environmentally Preferable
'Biobag indicator' autoclavable bags
  •  
    Material
    L×W
    Pk
  • Autoclavable bags, resistant to 121 °C , 'Biobag indicator' type 30
    Autoclavable bags, resistant to 121 °C , 'Biobag indicator' type 30
    129-0712
    Plastic
    300×500 mm
    500
    1 * 500 items
    1 * 500 items
    Specifications
    Environmentally Preferable
  • Autoclavable bags, resistant to 121 °C , 'Biobag indicator' type 60
    Autoclavable bags, resistant to 121 °C , 'Biobag indicator' type 60
    129-0713
    Plastic
    600×780 mm
    500
    1 * 500 items
    1 * 500 items
    Specifications
    Environmentally Preferable