Cassettes, Q Path® MacroStar VI
Supplier: VWR, part of Avantor
MacroStar VI
720-2181EA 47.34 USD
720-2181 720-2182 720-2176 720-2177 720-2174 720-2175 720-2178 720-2179 720-2180
Cassettes, Q Path® MacroStar VI
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with welded lid, supplied in dispensers for 500 cassettes. Packed in three boxes of 500 cassettes.
  • 30° angle
  • Writing area at the front and on both sides
  • Cassettes with ridges, which makes it easier to be opened after impregnation
詳細確認 About VWR