VWR® Q Path® MicroStar II, Cassettes, in Dispenser
Supplier: VWR
Q Path® MicroStar II
720-2382EA 211.62 USD
720-2382 720-2390 720-2385 720-2386 720-2383 720-2384 720-2389 720-2387 720-2388
VWR® Q Path® MicroStar II, Cassettes, in Dispenser
Embedding Cassettes Biopsy Processing Cassettes
Acetal polymer. Suitable for holding and identifying tissue samples in processing, embedding and sectioning procedures. Resistant to all commonly used histological solvents. Cassettes with tape (= rack) for Leica and Sakura cassette printers and separate disposable lid, supplied in racks of 40 cassettes.
  • Small opening for the filtration of fluids
  • Hinge and separate lid
  • 45° writing angle
詳細確認 About VWR