VWR®, Centrifuge Tubes, Light-Sensitive
Supplier: VWR
VWR®
525-0603EA 100.42 USD
525-0603 525-0602 525-0601 525-0600 525-0599 525-0598
VWR®, Centrifuge Tubes, Light-Sensitive
Tubes Centrifuge Tubes
PP, high grade gamma resistant, conical, amber, with white HDPE plug style cap with deep sealing area.
  • Tubes and caps are autoclavable and freezable
  • Gamma irradiated
  • Engraved graduations at the conical bottom of each tube
  • Graduations and writing area are chloroform-resistant
  • Longer length screw caps with sealing ring prevent any leakage
  • Available in 15 or 50 ml sizes and offered in a variety of packaging formats
  • Certified non cytotoxic, nuclease-free and pyrogen-free
  • Sustainable design based on virgin resins
  • Blocks 99% of light with wavelength less than 550 nm
  • During the freezing process, it is better to use plastic rack
  • During high-speed centrifugation, it is better to use suitable rotor adapters
Designed to provide protection for light-sensitive samples.
詳細確認 About VWR