Hydrogen peroxide 30% unstabilised, NORMATOM®, Ultrapure for trace metal analysis
Supplier: VWR Chemicals
|
Danger
|
85040.300EA
0
USD
85040.300
85040.270
Hydrogen peroxide 30% unstabilised, NORMATOM®, Ultrapure for trace metal analysis
Hydrogen peroxide
Formula:
H₂O₂ MW: 34.01 g/mol Boiling Pt: ~107 °C (1013 hPa) Melting Pt: <0 °C Density: 1.11 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011333 CAS Number: 7722-84-1 EINECS: 231-765-0 UN: 2014 ADR: 5.1 (8),II |
Specification Test Results
Assay | 30-32% |
Ag (Silver) | 10 ppt |
Al (Aluminium) | 50 ppt |
As (Arsenic) | 100 ppt |
Au (Gold) | 10 ppt |
B (Boron) | 100 ppt |
Ba (Barium) | 10 ppt |
Be (Beryllium) | 10 ppt |
Bi (Bismuth) | 10 ppt |
Ca (Calcium) | 100 ppt |
Cd (Cadmium) | 10 ppt |
Ce (Cerium) | 1 ppt |
Co (Cobalt) | 10 ppt |
Cr (Chromium) | 10 ppt |
Cs (Cesium) | 1 ppt |
Cu (Copper) | 10 ppt |
Dy (Dysprosium) | 1 ppt |
Er (Erbium) | 1 ppt |
Eu (Europium) | 1 ppt |
Fe (Iron) | 20 ppt |
Ga (Gallium) | 10 ppt |
Gd (Gadolinium) | 1 ppt |
Ge (Germanium) | 10 ppt |
Hf (Hafnium) | 1 ppt |
Hg (Mercury) | 50 ppt |
Ho (Holmium) | 1 ppt |
In (Indium) | 1 ppt |
K (Potassium) | 20 ppt |
La (Lanthanum) | 1 ppt |
Li (Lithium) | 10 ppt |
Lu (Lutetium) | 1 ppt |
Mg (Magnesium) | 20 ppt |
Mn (Manganese) | 10 ppt |
Mo (Molybdenum) | 10 ppt |
Na (Sodium) | 50 ppt |
Nb (Niobium) | 10 ppt |
Nd (Neodymium) | 1 ppt |
Ni (Nickel) | 20 ppt |
Pb (Lead) | 10 ppt |
Pd (Palladium) | 10 ppt |
Pr (Praseodymium) | 1 ppt |
Pt (Platinum) | 10 ppt |
Rb (Rubidium) | 10 ppt |
Re (Rhenium) | 10 ppt |
Rh (Rhodium) | 10 ppt |
Ru (Ruthenium) | 10 ppt |
Sb (Antimony) | 10 ppt |
Sc (Scandium) | 10 ppt |
Se (Selenium) | 100 ppt |
Sm (Samarium) | 1 ppt |
Sn (Tin) | 50 ppt |
Sr (Strontium) | 10 ppt |
Ta (Tantalum) | 10 ppt |
Tb (Terbium) | 1 ppt |
Te (Tellurium) | 1 ppt |
Th (Thorium) | 1 ppt |
Ti (Titanium) | 20 ppt |
Tl (Thallium) | 1 ppt |
Tm (Thulium) | 1 ppt |
U (Uranium) | 1 ppt |
V (Vanadium) | 10 ppt |
W (Tungsten) | 20 ppt |
Y (Yttrium) | 1 ppt |
Yb (Ytterbium) | 1 ppt |
Zn (Zinc) | 50 ppt |
Zr (Zirconium) | 10 ppt |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...