These sterile sleeve covers are made from a specially formulated breathable microporous fabric that provides significant fluid and barrier protection. The sleeves feature elastic openings and sonically welded seams.
- Gamma irradiated to a Sterility Assurance Level (SAL) of 10⁻⁶
- Lowest level of particle shedding: Level I Helmke Drum Classification
- Durable fabric with excellent breathability and water vapor transmission rate
- Significant fluid and particulate barrier: 99,9% bacterial filtration efficiency
- Soft, cloth-like fabric
Manufactured and packaged in an ISO Class 8 (Class 100000) cleanroom.
Tested and manufactured in an ISO certified facility under stringent process controls to ensure that each product meets exacting quality standards and performs to specification. Products are validated through independent lab testing.