VWR®, Microcentrifuge tubes, 1,7/2,0 ml
Supplier: VWR
VWRU87003-298EA 147.51 USD
VWRU87004-276 VWRU87003-298 VWRU87003-296 VWRU87003-294
VWR®, Microcentrifuge tubes, 1,7/2,0 ml
Tubes Microcentrifuge Tubes
PP, graduated, autoclavable and freezable with attached flat caps.
  • DNase-, RNase and endotoxin-free
  • Can withstand centrifugation to RCF 20000×g
  • Fit all standard and high capacity rotors
  • Tubes with X-Resin™ provides ultra-low binding characteristics
Medical grade superior clarity for visualisation of samples and isolated pellets. Graduated frosted side panel and flat snap cap provide surface for labelling.
  • DNase-, RNase and endotoxin-free
  • Can withstand centrifugation to RCF 20 000×g
  • Fit all standard and high capacity rotors
  • Tubes with X-Resin™ provides ultra-low binding characteristics
Medical grade superior clarity for visualisation of samples and isolated pellets. Graduated frosted side panel and flat snap cap provide surface for labelling.
詳細確認 About VWR