VWR®, Palladium standard solutions for ICP-MS, ARISTAR®
Supplier: Avantor
VWR®
457371KEA 493.02 USD
457371K 85587.180
VWR®, Palladium standard solutions for ICP-MS, ARISTAR®
Standards Cationic Standards Palladium Standards
  • Produced from high purity acids, water ASTM I 18 Megaohms
  • Solution assayed by titration
  • Final concentration verified against reference material from NIST
  • Total maximum uncertainty: 0.5% (typical)
  • Delivered with complete certificate of analysis
Certifications: Manufactured and tested in ISO 17034 / ISO 17025 accredited facilities
詳細確認 About VWR