VWR®, Colour-Coated Adhesion Slides
Supplier: VWR
VWR®
631-1176EA 0 USD
631-1176 631-1169 631-1175 631-1173 631-1172
VWR®, Colour-Coated Adhesion Slides
Slides Adhesive Slides
Microscope slides recommended for haematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
  • Smooth edges for operational safety
  • Sticky surface for stronger adhesion than with poly-l-lysine
  • Marking with conventional labelling system, pencils, marker pens and thermal transfer printers
Recommended for hematoxylin and eosin (H&E) stain, immunohistochemistry (IHC) and hybridisation on frozen sections in histopathology examination.
Packaging: 50 pieces per box. 50 boxes per case.
Caution: To be stored in dry and proper laboratory temperature conditions − shelf life of 60 months.
詳細確認 About VWR