VWR® Star Tec, ESD Chairs
Supplier: VWR
VWR®
132-1398EA 398.24 USD
132-1398 132-1399 132-1396 132-1397 132-1400 132-1401
VWR® Star Tec, ESD Chairs
Chairs
ESD chairs, with conductive seat and back, in black artificial leather, are soft and comfortable. The permanent backrest follows all movements and supports dynamic sitting in laboratories. Extra large, ergonomically-designed backrest with integrated lumbar support, the upholstery pads on the backrest provide maximum seating comfort and an optimum micro climate. The tapered backrest, in the upper area, offers elbow room for flexible work in laboratories and maximum freedom of movement.
  • Reliable ESD protection
  • Washable, resistant to desinfectants
  • Robust and easy to clean
  • Backrest adjustable in height and inclination
  • Seat inclination adjustment supports a forward-angled working position, which is typical for laboratory work
  • Every function can be operated comfortably and easily in a sitting position
  • Pratical handle on the backrest provides maximum mobility and easy handling
  • Polished aluminium star base
Certifications: Suitable for use in biosafety laboratories BSL 1 and BSL 2, GMP conformity. Compliant to ESD norm EN 61340-5-1.
詳細確認 About VWR