Dual-cooled vertical electrophoresis systems, kuroGEL Verti 1816K and 1824K
700-0189EA 0 USD
VWRI700-0189
Dual-cooled vertical electrophoresis systems, kuroGEL Verti 1816K and 1824K
The kuroGEL Verti dual-cooled units are available in standard (18×16 cm) or tall (18×24 cm) formats.
  • Produce straight lanes and sharp, well defined bands
  • Run up to four gels (maximum 112 samples) at one time under identical conditions
  • Run gels at uniform temperature from 1 to 45 °C
  • Accommodates denaturing and native polyacrylamide gels and 2-D electrophoresis
  • A wide array of accessories allow gel configurations to be tailored to meet individual needs
Delivery information: Complete system includes lower and upper buffer chambers; safety lid with high voltage leads; heat exchanger; glass plates 18×16 cm for Verti 1816K or 18×24 cm for Verti 1824K (6 pcs); clamp assemblies, 16 cm (4 pcs) for Verti 1816K or 8 cm (4 pcs) and 16 cm (4 pcs) for Verti 1824K; cams (6 pcs); slotted gaskets for upper buffer chamber (2 pcs); dual gel casting stand with levelling base; bubble level; laminated gaskets for caster (2 pcs); buffer dam; alignment template; plate separation tool; gel seal; combs, 15 well, 1,5 mm thick (2 pcs); spacers, 1,5 mm thick, 2 cm wide (4 pcs).
詳細確認 About VWR