Glycine ≥99.5%, Electran for molecular biology

Synonyms: Aminoacetic acidH-Gly-OH

444495DEA 111.38 USD
444495D
Glycine ≥99.5%, Electran for molecular biology
Glycine
Formula: NH₂CH₂COOH
MW: 75.07 g/mol
Boiling Pt: 181 °C (1013 hPa)
Melting Pt: 83 °C
Density: 1.64 g/cm³ (20 °C)
Storage Temperature: Ambient
MDL Number: MFCD00008131
CAS Number: 56-40-6
EINECS: 200-272-2
Merck Index: 13,04504
https://jp.vwr.com/stibo/web/std.lang.all/19/10/2301910.jpg

Order Now

Specification Test Results

Assay Min. 99.5 %
Appearance White crystalline powder
Identity (IR) Passes test
DNases Not detected
RNases Not detected
Proteases Not detected
Absorbance (260 nm) (0.1 mol/l) Max. 0.010
Absorbance (280 nm) (0.1 mol/l) Max. 0.010
Heavy metals (as Pb) Max. 0.001 %
Loss on drying Max. 0.10 %
pH (5 %) 5.9 - 6.4
NH4 (Ammonium) Max. 0.0200 %
SO4 (Sulphate) Max. 0.005 %
As (Arsenic) Max. 0.0001 %
Cl (Chloride) Max. 0.004 %
Fe (Iron) Max. 0.0005 %
Pb (Lead) Max. 0.0005 %
Pb (Lead) Max. 0.0005 %

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

詳細確認 About VWR