IPTG (isopropyl-β-D-thiogalactopyranoside) ≥98% (by HPLC on dried basis), Biotech Reagent, cGMP-manufactured, J.T.Baker®
Supplier: AVANTOR PERFORMANCE MATERIALS
Warning
|
Synonyms:
(2R,3R,4S,5R,6S)-2-(Hydroxymethyl)-6-(isopropylthio)tetrahydro-2H-pyran-3,4,5-triol
, Isopropyl Beta-D-1-thiogalactopyranoside
Reduce risk in your bioprocessing applications with Avantor J.T.Baker brand cGMP manufactured IPTG.
- cGMP manufactured under ICHQ7
- High purity (>98% by HPLC)
- Low microbial, trace metals and endotoxin content
- Consistent lot-to-lot protein induction performance
- TSE/BSE-free
- 1,4-Dioxane-free
J.T.Baker brand cGMP manufactured IPTG is performance tested to assure consistent lot-to-lot biological induction, and processed to remove and control product/process related impurities, with minimal lot-to-lot variability for improved protein expression.
Fully validated and documented through Batch Process Records (BPR) including analytical test data, stability protocol and impurity profile.
Packaging: Packed in moisture permeation resistant controlled packaging.
Formula:
C₉H₁₈O₅S MW: 238.31 g/mol Melting Pt: 110…122 °C |
MDL Number:
MFCD00063273 CAS Number: 367-93-1 |
Specification Test Results
GMP Manufactured Product | |
Store and ship product between 2 and 8 °C | |
CAUTION: For Manufacturing, processing or repackaging | |
Appearance (white to off-white solid) | Passes Test |
Identification (IR) | Passes Test |
pH of 5% Solution | 5.00 - 7.00 |
Specific Rotation @ 25 °C | -34.5 - -28.5 Degree |
Melting Point | 110 - 122 °C |
Water (H₂O), For Information Only | |
Purity (by HPLC) (anhydrous basis) | ≥98 % |
Assay (by HPLC) (anhydrous basis) | 98 - 102 |
Endotoxin Concentration, EU/g | |
Residual Methanol | ≤3000 ppm |
Residual Ethanol | ≤5000 ppm |
Residual Isopropanol | ≤5000 ppm |
Residual Ethyl Acetate | ≤5000 ppm |
Total Aerobic Microbial Count | ≤100 cfu/g |
Total Yeast and Mold Count | ≤100 cfu/g |
Biological Induction Characterization (induction response @ 1mM) | |
Arsenic (As), For Information Only | |
Cadmium (Cd), For Information Only | |
Mercury (Hg), For Information Only | |
Lead (Pb), For Information Only | |
Cobalt (Co), For Information Only | |
Nickel (Ni), For Information Only | |
Vanadium (V), For Information Only | |
Silver (Ag), For Information Only | |
Gold (Au), For Information Only | |
Iridium (Ir), For Information Only | |
Osmium (Os), For Information Only | |
Palladium (Pd), For Information Only | |
Platinum (Pt), For Information Only | |
Rhodium (Rh), For Information Only | |
Ruthenium (Ru), For Information Only | |
Selenium (Se), For Information Only | |
Thallium (Tl), For Information Only | |
Barium (Ba), For Information Only | |
Chromium (Cr), For Information Only | |
Copper (Cu), For Information Only | |
Lithium (Li), For Information Only | |
Molybdenum (Mo), For Information Only | |
Antimony (Sb), For Information Only | |
Tin (Sn), For Information Only | |
Aluminum (Al), For Information Only | |
Iron (Fe), For Information Only | |
Manganese (Mn), For Information Only | |
Zinc (Zn), For Information Only | |
Residual Solvents: The Class 3 solvents Ethanol, Isopropanol, Ethyl | |
Acetate and the Class 2 solvent Methanol are likely to be present. These solvents are quantified and reported on each batch. No other | |
Class 1,2,3, or other solvents are used or produced in the production | |
process. Dioxane was not used in the manufacturing process. | |
Bacterial Induction characterization: Test performed at 1 mM of IPTG (E. | |
Coli. protein expression) | |
IPTG is highly hygroscopic. Allow packaging/container to equilibrate to | |
room temperature before opening for analysis. Ensure sample is kept dry | |
(such as in a desiccator) after opening. |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...