VWR® pHenomenal® IDP pH Electrodes
Supplier: VWR
VWR® PHenomenal®
662-2298EA 754.12 USD
662-2298 662-2297 662-2296
VWR® pHenomenal® IDP pH Electrodes
Electrodes pH Electrodes
These IDP (Intelligent Digital Probe) pH/temperature electrodes can be used with the pHenomenal® MD 8000 H and MD 8000 L multi-parameter meters.
  • Calibration data (serial number, calibration status) is stored in the electrode/probe, transmitted to the meter and displayed
  • Proven reliability and accuracy
  • Correct measurement results due to direct signal processing in the sensor and digital transmission of values
  • All parameters are easily measured with cable lengths of up to 100 metres
  • Same universal connector for all electrodes and probes
Temperature probe: NTC 30 kOhm
The electrodes can provide the following data shown on the meter's screen:
- Model name
- Serial number
- Calibration status (+++, ++, +, –, blank)
- Calibration history includes the last 10 calibrations, including buffers used, mV values, etc.
Delivery information: Includes digital pH electrode only.
詳細確認 About VWR