VWR®, Shoe covers, non-skid, irradiated
Supplier: VWR
VWR®
VWRU414004-475EA 41.47 USD
VWRU414004-475 VWRU414004-476 VWRU414004-479 VWRU414004-477 VWRU414004-478
VWR®, Shoe covers, non-skid, irradiated
Disposable Garments
Irradiated non-skid shoe covers are manufactured from a impervious, clean and low-linting fabric.
  • Superior coefficient of friction
  • Non-skid soles provide maximum traction on wet and smooth surfaces
  • Gamma irradiated to a Sterility Assurance Level (SAL) of 10-4
Engineered to optimise skid resistance, barrier protection, cleanliness, and comfort.
Certifications: Tested and manufactured in an ISO certified facility under stringent process controls. Lot-controlled and validated through independent laboratory testing. Manufactured in an ISO Class 8 (FED-STD-209E Class 100000/M6.5) cleanroom.
Packaging: Bulk packed in sealed PE bags contained within a larger carton liner. Each case is supplied with a certificate of irradiation.
詳細確認 About VWR