VWR®, LTS® Compatible Filter Tips
Supplier: VWR
VWR®
732-3639EA 136.28 USD
732-3639 732-3640 732-3641 732-3642 732-3643
VWR®, LTS® Compatible Filter Tips
Pipette Tips
VWR® LTS® compatible filter tips feature a flexible seal area and a positive stop resulting in a secure seal with light ejection force on LTS® compatible pipettors.
  • Compatible with the LTS® style pipettors
  • Maximises sample recovery with low binding resin and a fine point design
  • Improves ergonomics by reducing the force required to attach and eject a tip
  • Autoclavable at 1 bar, 122 °C for 15 minutes
  • Manufactured under an ISO 9001 quality management system
VWR® tips use a low retention resin that is up to ten times more hydrophobic than other polypropylene pipette tips, enabling viscous samples like protein and DNA solutions to dispense easily without collecting inside the tip.
Certifications: Certified RNase-, DNase-, endotoxin-, metal-, and human DNA-free.
詳細確認 About VWR