Dimethyldichlorosilane 2% in heptane, Electran for electrophoresis
Supplier: VWR Chemicals
|
Danger
|
Synonyms:
Dichlorodimethylsilane
, Dichloro(dimethyl)silane
437494JEA
237.6
USD
437494J
Dimethyldichlorosilane 2% in heptane, Electran for electrophoresis
Dimethyldichlorosilane
Designed to give water repellant properties to glassware. Suitable for use in electrophoresis to prevent adhesion of gels to glass plates.
Formula:
C₂H₆Cl₂Si MW: 129.06 g/mol |
MDL Number:
MFCD00000491 CAS Number: 75-78-5 EINECS: 200-901-0 UN: 1993 ADR: 3,II |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...