4-Aminoantipyrine, TECHNICAL
Supplier: VWR Chemicals
|
Warning
|
Synonyms:
4-Amino-2,3-dimethyl-1-phenyl-3-pyrazolin-5-one
, 4-Aminophenazone
21160.185EA
404.66
USD
21160.185
4-Aminoantipyrine, TECHNICAL
4-Aminoantipyrine
Formula:
C₁₁H₁₃N₃O MW: 203.24 g/mol Melting Pt: 107…109 °C Storage Temperature: Ambient |
MDL Number:
MFCD00003145 CAS Number: 83-07-8 EINECS: 201-452-3 Merck Index: 13,00596 |
Specification Test Results
Identification | Passes test |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...