VWR® Professional Magnetic Stirrers
Supplier: VWR
VWR® Professional
444-0611EA 1259.39 USD
VWRU97042-750 444-0611 VWRU97042-710 444-0616 444-0613 VWRU97042-706 444-0612 444-0615 444-0614
VWR® Professional Magnetic Stirrers
Stirrers Magnetic Stirrers
VWR® Professional magnetic stirrers are designed for applications that require exceptional accuracy, stability and repeatability. Stirring function, with continuous duty motor and powerful magnet, maintains set speed even under changing load or viscosity. Touch pad controls with easy to read, independent LED displays for speed, and time, allow the operator to view all settings at once.
  • Microprocessor control
  • Separate digital displays for speed and time. Display will show last used settings, even after power has been turned off
  • Chemically resistant housing
  • Ceramic top plate
Rear housing features a built-in support rod holder with locking knob. Low profile design takes up less space and fits into fume hoods. Spill resistant housing channels fluids away from internal components. An audible alarm will sound when time reaches zero in timed mode. Microprocessor control with enhanced electronics regulates stirring - speed control is precise and stirring consistent at all speeds. Ramping feature slowly increases speed for improved safety and enhanced coupling. Avoids splashing, improves spin bar control and provides excellent low end speed control. The ceramic tops feature a chemically resistant, reflective white top plate surface that is easy to clean.
Delivery information: Each unit is supplied complete with a PTFE coated magnetic stirring bar.
詳細確認 About VWR