VWR®, Disposable Spatulas
Supplier: VWR
VWR®
231-0416EA 0 USD
231-0416 231-0106 231-0104 231-0369 231-0105 231-0103 231-0593
VWR®, Disposable Spatulas
Spatulas
PP. Multi-purpose, robust, inexpensive tools for collecting, transferring and processing samples of chemical and biological materials.
  • Resistant to dilute acids and alkalis
  • Reduces cross-contamination
  • Autoclavable
The innovative shape of these spatulas allows them to be used as a spatula, spoon, scoop, scraper, mixing rod or for wiping. Ideal for recovering samples that have been frozen in liquid nitrogen. The macro type disposable spatula is 50% larger than the standard disposable spatula. The long shaft makes it easier to transfer material from larger containers.
Certifications: Compliant with FDA standards for applications involving contact with food.
詳細確認 About VWR