Hydrochloric acid 32%, NORMATOM®, Ultrapure for trace metal analysis
Supplier: VWR Chemicals
|
Danger
|
Synonyms:
Muriatic acid
83878.300EA
2751.89
USD
83878.300
83878.230
83878.270
83878.290
Hydrochloric acid 32%, NORMATOM®, Ultrapure for trace metal analysis
Hydrochloric acid
Formula:
HCl MW: 36.46 g/mol Boiling Pt: 110 °C (1013 hPa) Melting Pt: –30 °C Density: 1.18 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011324 CAS Number: 7647-01-0 EINECS: 231-595-7 UN: 1789 ADR: 8,II REACH: 01-2119484862-27 |
Specification Test Results
Assay (HCl, w/w) | 32 - 35% |
Ag (Silver) | max. 10 ppt |
Al (Aluminium) | max. 20 ppt |
As (Arsenic) | max. 50 ppt |
Au (Gold) | max. 50 ppt |
B (Boron) | max. 100 ppt |
Ba (Barium) | max. 10 ppt |
Be (Beryllium) | max. 10 ppt |
Bi (Bismuth) | max. 10 ppt |
Ca (Calcium) | max. 10 ppt |
Cd (Cadmium) | max. 10 ppt |
Ce (Cerium) | max. 10 ppt |
Co (Cobalt) | max. 10 ppt |
Cr (Chromium) | max. 10 ppt |
Cs (Cesium) | max. 10 ppt |
Cu (Copper) | max. 10 ppt |
Dy (Dysprosium) | max. 1 ppt |
Er (Erbium) | max. 1 ppt |
Eu (Europium) | max. 1 ppt |
Fe (Iron) | max. 10 ppt |
Ga (Gallium) | max. 10 ppt |
Gd (Gadolinium) | max. 1 ppt |
Hf (Hafnium) | max. 10 ppt |
Hg (Mercury) | max. 50 ppt |
Ho (Holmium) | max. 1 ppt |
In (Indium) | max. 1 ppt |
K (Potassium) | max. 10 ppt |
La (Lanthanum) | max. 1 ppt |
Li (Lithium) | max. 10 ppt |
Lu (Lutetium) | max. 10 ppt |
Mg (Magnesium) | max. 10 ppt |
Mn (Manganese) | max. 10 ppt |
Mo (Molybdenum) | max. 10 ppt |
Na (Sodium) | max. 10 ppt |
Nb (Niobium) | max. 1 ppt |
Nd (Neodymium) | max. 1 ppt |
Ni (Nickel) | max. 20 ppt |
Pb (Lead) | max. 10 ppt |
Pd (Palladium) | Information Only |
Pr (Praseodymium) | max. 1 ppt |
Pt (Platinum) | Information Only |
Rb (Rubidium) | max. 10 ppt |
Re (Rhenium) | max. 10 ppt |
Rh (Rhodium) | max. 10 ppt |
Ru (Ruthenium) | max. 10 ppt |
Sb (Antimony) | max. 20 ppt |
Sc (Scandium) | max. 10 ppt |
Se (Selenium) | Information Only |
Sm (Samarium) | max. 1 ppt |
Sn (Tin) | max. 20 ppt |
Sr (Strontium) | max. 10 ppt |
Ta (Tantalum) | Information Only |
Tb (Terbium) | max. 1 ppt |
Te (Tellurium) | max. 1 ppt |
Th (Thorium) | max. 1 ppt |
Ti (Titanium) | max. 10 ppt |
Tl (Thallium) | max. 10 ppt |
Tm (Thulium) | max. 1 ppt |
U (Uranium) | max. 1 ppt |
V (Vanadium) | max. 10 ppt |
W (Tungsten) | max. 10 ppt |
Y (Yttrium) | max. 1 ppt |
Yb (Ytterbium) | max. 1 ppt |
Zn (Zinc) | max. 10 ppt |
Zr (Zirconium) | max. 10 ppt |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...