Nitric acid 67%, NORMATOM®, Ultrapure for trace metal analysis
Supplier: VWR Chemicals
|
Danger
|
83879.300EA
2788.53
USD
83879.300
83879.230
83879.270
83879.290
Nitric acid 67%, NORMATOM®, Ultrapure for trace metal analysis
Nitric acid
Formula:
HNO₃ MW: 63.01 g/mol Boiling Pt: 120.5 °C (1013 hPa) Melting Pt: –42 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 EINECS: 231-714-2 UN: 2031 ADR: 8 (5.1),II Merck Index: 13,06608 |
Specification Test Results
Assay | Min. 67 % |
Ag (Silver) | Max. 10 ppt |
Al (Aluminium) | Max. 20 ppt |
As (Arsenic) | Max. 20 ppt |
Au (Gold) | Max. 20 ppt |
B (Boron) | Max. 20 ppt |
Ba (Barium) | Max. 10 ppt |
Be (Beryllium) | Max. 10 ppt |
Bi (Bismuth) | Max. 10 ppt |
Ca (Calcium) | Max. 20 ppt |
Cd (Cadmium) | Max. 10 ppt |
Ce (Cerium) | Max. 10 ppt |
Co (Cobalt) | Max. 10 ppt |
Cr (Chromium) | Max. 20 ppt |
Cs (Cesium) | Max. 10 ppt |
Cu (Copper) | Max. 20 ppt |
Dy (Dysprosium) | Max. 1 ppt |
Er (Erbium) | Max. 1 ppt |
Eu (Europium) | Max. 1 ppt |
Fe (Iron) | Max. 20 ppt |
Ga (Gallium) | Max. 10 ppt |
Gd (Gadolinium) | Max. 1 ppt |
Ge (Germanium) | Max. 10 ppt |
Hf (Hafnium) | Max. 10 ppt |
Hg (Mercury) | Max. 100 ppt |
Ho (Holmium) | Max. 1 ppt |
In (Indium) | Max. 1 ppt |
K (Potassium) | Max. 10 ppt |
La (Lanthanum) | Max. 1 ppt |
Li (Lithium) | Max. 10 ppt |
Lu (Lutetium) | Max. 1 ppt |
Mg (Magnesium) | Max. 10 ppt |
Mn (Manganese) | Max. 10 ppt |
Mo (Molybdenum) | Max. 10 ppt |
Na (Sodium) | Max. 10 ppt |
Nb (Niobium) | Max. 1 ppt |
Nd (Neodymium) | Max. 1 ppt |
Ni (Nickel) | Max. 50 ppt |
Pb (Lead) | Max. 10 ppt |
Pd (Palladium) | Max. 20 ppt |
Pr (Praseodymium) | Max. 1 ppt |
Pt (Platinum) | Max. 20 ppt |
Rb (Rubidium) | Max. 10 ppt |
Re (Rhenium) | Max. 10 ppt |
Rh (Rhodium) | Max 10 ppt |
Ru (Ruthenium) | Max. 20 ppt |
Sb (Antimony) | Max. 10 ppt |
Sc (Scandium) | Max. 10 ppt |
Sm (Samarium) | Max. 1 ppt |
Sn (Tin) | Max. 20 ppt |
Sr (Strontium) | Max. 10 ppt |
Tb (Terbium) | Max. 1 ppt |
Te (Tellurium) | Max. 1 ppt |
Th (Thorium) | Max. 1 ppt |
Ti (Titanium) | Max. 10 ppt |
Tl (Thallium) | Max. 10 ppt |
Tm (Thulium) | Max. 1 ppt |
U (Uranium) | Max. 1 ppt |
V (Vanadium) | Max. 10 ppt |
W (Tungsten) | Max. 10 ppt |
Y (Yttrium) | Max. 1 ppt |
Yb (Ytterbium) | Max. 1 ppt |
Zn (Zinc) | Max. 20 ppt |
Zr (Zirconium) | Max. 10 ppt |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...