Perchloric acid 65%, NORMATOM® for trace metal analysis
Supplier: VWR Chemicals
|
Danger
|
83874.320EA
680.91
USD
83874.320
83874.270
Perchloric acid 65%, NORMATOM® for trace metal analysis
Perchloric acid
Formula:
HClO₄ MW: 100.46 g/mol Boiling Pt: 160…181 °C (1013 hPa) Melting Pt: –112,2 °C Density: 1.53…1.768 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011325 CAS Number: 7601-90-3 EINECS: 231-512-4 UN: 1873 ADR: 5.1 (8),I |
Specification Test Results
Assay | 65 - 71 % |
Colouration | Max. 10 APHA |
Free chlorine | Max. 0.5 ppm |
Ignition residue (SO₄) | Max. 30 ppm |
Total N (Nitrogen) | Max. 10 ppm |
Cl (Chloride) | Max. 3 ppm |
ClO₃ (Chlorate) | Max. 10 ppm |
PO₄+ SiO₄(as SiO₂) | Max. 5 ppm |
SO4 (Sulphate) | Max. 10 ppm |
Ag (Silver) | Max. 1 ppb |
Al (Aluminium) | Max. 1 ppb |
As (Arsenic) | Max. 0.5 ppb |
Au (Gold) | Max. 0.5 ppb |
Ba (Barium) | Max. 1 ppb |
Be (Beryllium) | Max. 0.5 ppb |
Bi (Bismuth) | Max. 0.5 ppb |
Ca (Calcium) | Max. 1 ppb |
Cd (Cadmium) | Max. 1 ppb |
Ce (Cerium) | Max. 0.5 ppb |
Co (Cobalt) | Max. 0.5 ppb |
Cs (Cesium) | Max. 0.5 ppb |
Cu (Copper) | Max. 0.5 ppb |
Dy (Dysprosium) | Max. 0.5 ppb |
Er (Erbium) | Max. 0.5 ppb |
Eu (Europium) | Max. 0.5 ppb |
Fe (Iron) | Max. 1 ppb |
Ga (Gallium) | Max. 0.5 ppb |
Gd (Gadolinium) | Max. 0.5 ppb |
Ho (Holmium) | Max. 0.5 ppb |
In (Indium) | Max. 0.5 ppb |
K (Potassium) | Max. 1 ppb |
La (Lanthanum) | Max. 0.5 ppb |
Li (Lithium) | Max. 0.5 ppb |
Lu (Lutetium) | Max. 0.5 ppb |
Mg (Magnesium) | Max. 1 ppb |
Mn (Manganese) | Max. 1 ppb |
Mo (Molybdenum) | Max. 0.5 ppb |
Na (Sodium) | Max. 1 ppb |
Nd (Neodymium) | Max. 0.5 ppb |
Ni (Nickel) | Max. 1 ppb |
Pb (Lead) | Max. 1 ppb |
Pd (Palladium) | Max. 0.5 ppb |
Pr (Praseodymium) | Max. 0.5 ppb |
Pt (Platinum) | Max. 0.5 ppb |
Rb (Rubidium) | Max. 0.5 ppb |
Rh (Rhodium) | Max. 0.5 ppb |
Ru (Ruthenium) | Max. 0.5 ppb |
Sb (Antimony) | Max. 0.5 ppb |
Sc (Scandium) | Max. 0.5 ppb |
Sm (Samarium) | Max. 0.5 ppb |
Sn (Tin) | Max. 1 ppb |
Sr (Strontium) | Max. 0.5 ppb |
Tb (Terbium) | Max. 0.5 ppb |
Te (Tellurium) | Max. 0.5 ppb |
Th (Thorium) | Max. 1 ppb |
Ti (Titanium) | Max. 1 ppb |
Tl (Thallium) | Max. 0.5 ppb |
Tm (Thulium) | Max. 0.5 ppb |
U (Uranium) | Max. 0.5 ppb |
V (Vanadium) | Max. 0.5 ppb |
Y (Yttrium) | Max. 0.5 ppb |
Yb (Ytterbium) | Max. 0.5 ppb |
Zn (Zinc) | Max. 1 ppb |
Zr (Zirconium) | Max. 0.5 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...