Nitric acid 67%, NORMATOM® for trace metal analysis
Supplier: VWR Chemicals
|
Danger
|
83872.290EA
421
USD
83872.290
83872.270
83872.330
Nitric acid 67%, NORMATOM® for trace metal analysis
Nitric acid
Formula:
HNO₃ MW: 63.01 g/mol Boiling Pt: 120.5 °C (1013 hPa) Melting Pt: –42 °C Density: 1.4 g/cm³ (20 °C) Storage Temperature: Ambient |
MDL Number:
MFCD00011349 EINECS: 231-714-2 UN: 2031 ADR: 8 (5.1),II Merck Index: 13,06608 |
Specification Test Results
Assay | 67 - 69 % |
Colouration | Max. 10 APHA |
Cl (Chloride) | Max. 0.2 ppm |
Total P (Phosphorus) | Max. 0.01 ppm |
Total S (Sulphur) | Max. 0.3 ppm |
Ag (Silver) | Max. 0.1 ppb |
Al (Aluminium) | Max. 1 ppb |
As (Arsenic) | Max. 0.5 ppb |
Au (Gold) | Max. 0.1 ppb |
B (Boron) | Max. 1 ppb |
Ba (Barium) | Max. 0.1 ppb |
Be (Beryllium) | Max. 0.1 ppb |
Bi (Bismuth) | Max. 0.1 ppb |
Ca (Calcium) | Max. 1 ppb |
Cd (Cadmium) | Max. 0.5 ppb |
Ce (Cerium) | Max. 0.1 ppb |
Co (Cobalt) | Max. 0.5 ppb |
Cr (Chromium) | Max. 1 ppb |
Cs (Cesium) | Max. 0.1 ppb |
Cu (Copper) | Max. 0.5 ppb |
Dy (Dysprosium) | Max. 0.1 ppb |
Er (Erbium) | Max. 0.1 ppb |
Eu (Europium) | Max. 0.1 ppb |
Fe (Iron) | Max. 1 ppb |
Ga (Gallium) | Max. 0.1 ppb |
Gd (Gadolinium) | Max. 0.1 ppb |
Ge (Germanium) | Max. 0.1 ppb |
Hf (Hafnium) | Max. 0.1 ppb |
Hg (Mercury) | Max. 0.1 ppb |
Ho (Holmium) | Max. 0.1 ppb |
In (Indium) | Max. 0.1 ppb |
K (Potassium) | Max. 1 ppb |
La (Lanthanum) | Max. 0.1 ppb |
Li (Lithium) | Max. 0.1 ppb |
Lu (Lutetium) | Max. 0.1 ppb |
Mg (Magnesium) | Max. 1 ppb |
Mn (Manganese) | Max. 0.1 ppb |
Mo (Molybdenum) | Max. 0.1 ppb |
Na (Sodium) | Max. 1 ppb |
Nb (Niobium) | Max. 0.1 ppb |
Nd (Neodymium) | Max. 0.1 ppb |
Ni (Nickel) | Max. 0.5 ppb |
Pb (Lead) | Max. 0.1 ppb |
Pd (Palladium) | Max. 0.5 ppb |
Pr (Praseodymium) | Max. 0.1 ppb |
Pt (Platinum) | Max. 0.5 ppb |
Rb (Rubidium) | Max. 0.1 ppb |
Re (Rhenium) | Max. 0.1 ppb |
Rh (Rhodium) | Max. 0.5 ppb |
Ru (Ruthenium) | Max. 0.500 ppb |
Sb (Antimony) | Max. 0.500 ppb |
Sc (Scandium) | Max. 0.100 ppb |
Se (Selenium) | Max. 1 ppb |
Sm (Samarium) | Max. 0.100 ppb |
Sn (Stagno) | Max. 0,500 ppb |
Sr (Strontium) | Max. 0.100 ppb |
Tb (Terbium) | Max. 0.100 ppb |
Te (Tellurium) | Max. 0.100 ppb |
Th (Thorium) | Max. 0.100 ppb |
Ti (Titanium) | Max. 0.500 ppb |
Tl (Thallium) | Max. 0.100 ppb |
Tm (Thulium) | Max. 0.100 ppb |
U (Uranium) | Max. 0.100 ppb |
V (Vanadium) | Max. 0.500 ppb |
W (Tungsten) | Max. 0.100 ppb |
Y (Yttrium) | Max. 0.100 ppb |
Yb (Ytterbium) | Max. 0.100 ppb |
Zn (Zinc) | Max. 0.500 ppb |
Zr (Zirconium) | Max. 0.100 ppb |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...