n-Octane ≥95%, TECHNICAL
Supplier: VWR Chemicals
|
Danger
|
26024.292EA
76.51
USD
26024.292
n-Octane ≥95%, TECHNICAL
n-Octane
Formula:
H₃C(CH₂)₆CH₃ MW: 114.23 g/mol Boiling Pt: 126 °C (1013 hPa) Melting Pt: –57 °C Density: 0.703 g/cm³ (20 °C) Flash Pt: 8 °C Storage Temperature: Ambient |
MDL Number:
MFCD00009556 CAS Number: 111-65-9 EINECS: 203-892-1 UN: 1262 ADR: 3,II Merck Index: 12,06847 |
Specification Test Results
Assay | Min. 95 % |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...