VWR®, Bottle-Top Vacuum Filtration Systems, High Performance
Supplier: VWR
VWR®
514-0359EA 744.79 USD
514-0359 514-0317 514-0318 514-0355 514-0358 514-0296 514-0297 514-0298 514-0354 514-0299 514-0350 514-0308 514-0309 514-0304 514-0348 514-0305 514-0349 514-0306 514-0307 514-0366 514-0300 514-0367 514-0301 514-0302 514-0303 514-0362 514-0363 514-0364 514-0360
VWR®, Bottle-Top Vacuum Filtration Systems, High Performance
Filters Bottle Top Filters
Designed for the preparation of buffers, tissue culture media, microbiological media and other biological fluids. These vacuum filtration systems feature large diameter high performance PES membranes for guaranteed performance and maximum throughput. The full assembly includes a filtration unit and a receiver bottle with an ergonomic shape, moulded finger grips and a patented bottle cap. The centrifuge tube and funnel system allows the user to filter fluids directly into a 15 or 50 ml tube.
  • Quick-Connect™ pedestal base or cradle ring allows for hands-free filtration Weighted for extra stability
  • Quick-Connect™ pedestal base ensures that the unit is always correctly placed
  • Forward-facing, large, raised graduation marks allow for easy visualisation of volumes
  • Vacuum on/off switch located at the front
  • Vacuum hose adapter for use with conventional filtration set-ups
Each separate piece is compatible with the Quick-Connect™ pedestal base providing a convenient on/off vacuum flow control, which allows for a convenient single-use vacuum hose connection and one handed operation. A cradle ring is also available for use with existing laboratory stands. Systems are available with 0,1 µm (mycoplasma filtering grade), 0,2 µm (sterilising grade) or 0,45 µm (clarification grade) asymmetric PES membranes. All items are made from FDA grade, Class VI materials and are gamma sterilised VD Max 10⁻⁶ in a 100000 cleanroom in an ISO 13485 standard facility.
You can choose between complete systems and filtration funnels only. For the complete systems there is a choice of receivers: 15 or 50 ml centrifuge tubes; 250, 500 or 1000 ml media bottles. The 250, 500 and 1000 ml filtration funnels have a GL 45 thread.
Delivery information: Complete system includes filtration funnel (PS), media bottle (PS) or centrifuge tube (PP) and hose adapter.
詳細確認 About VWR