VWR® AVS TITRINORM, Yttrium Standard Solution, 1000 mg/L Y in dil. Nitric Acid, Standard for AAS
Supplier: Avantor
VWR® AVS TITRINORM
86723.180EA
99.5
USD
86723.180
86723.260
VWR® AVS TITRINORM, Yttrium Standard Solution, 1000 mg/L Y in dil. Nitric Acid, Standard for AAS
Standards
Cationic Standards
Yttrium Standards